

2,4,6-Trimethylphenylhydrazine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76195-82-9
:
Formula:C₉H₁₅ClN₂
MDL Number:MFCD00052268
:186.69 g/mol
Melting Pt:∼, 185 °C (dec.)
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together