2-(1-Cyclohexenyl)cyclohexanone ≥85% cont. ca 10% 2-cyclohexylidenecyclohexanone
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1502-22-3
:
Formula:C₁₂H₁₈O
MDL Number:MFCD00013796
:178.28 g/mol
Boiling Pt:265 °C
Density:0,998
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:216-120-3
Related Information
Frequently Bought Together