Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Hydroxy-4-methyl-8-nitrocoumarin
7-Hydroxy-4-methyl-8-nitrocoumarin 98%
7-Hydroxy-4-methyl-8-nitrocoumarin 98%
7-Hydroxy-4-methyl-8-nitrocoumarin 98%
L07896.03
:
Thermo Fisher Scientific
:
7-Hydroxy-4-methyl-8-nitrocoumarin 98%
L07896.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
19037-69-5
:
:
C₁₀H₇NO₅
:
MFCD00051824
:
221.17 g/mol
:
∼ 255 °C (dec.)
:
GHS07
:
Ambient