
5-Chlorothiazol-2-amine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55506-37-1
:
Formula:C3H4Cl2N2S
MDL Number:MFCD00039006
:171.05 g/mol
Melting Pt:168…172 °C
EINECS:259-680-4
:GHS06
Storage Temperature:Ambient
Related Information
Frequently Bought Together