

N,N'-Dibenzylethylenediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-28-3
:
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00004771
:240.35 g/mol
Merck Index:14,1064
Boiling Pt:184…186 °C (2 mmHg)
Melting Pt:23…25 °C
Density:1,025
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:205-408-4
UN:2735
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together