N'-Ethyl-N,N-dimethylethylenediamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-83-1
:
Formula:CH₃CH₂NHCH₂CH₂N(CH₃)₂
MDL Number:MFCD00009034
:116.21 g/mol
Boiling Pt:134…135 °C
Density:0,818
Flash Pt:23 °C (73 °F)
Storage Temperature:Ambient
EINECS:204-656-0
ADR:8,II
:GHS05
:GHS02
Related Information
Frequently Bought Together