
1,2-Bis(chlorodimethylsilyl)ethane 90%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13528-93-3
:
Formula:C₆H₁₆Cl₂Si₂
MDL Number:MFCD00000505
:215.26 g/mol
Boiling Pt:198…200 °C
Melting Pt:34…40 °C
Flash Pt:65 °C (149 °F)
Storage Temperature:Ambient
EINECS:236-871-0
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together