5-Nitrothiophene-2-carboxaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4521-33-9
:
Formula:C5H3NO3S
MDL Number:MFCD00005433
:157.15 g/mol
Melting Pt:72…76 °C
EINECS:224-850-9
Related Information
Frequently Bought Together