
Tetraphenylphosphine imide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2325-27-1
:
Formula:C₂₄H₂₀NP
MDL Number:MFCD00003044
:353.405 g/mol
Melting Pt:128…132 °C
EINECS:219-039-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together