Order Entry
Northern Ireland
ContactUsLinkComponent
1-(trans-Cinnamoyl)imidazole 98%
Irritant
1-(trans-Cinnamoyl)imidazole 98%
Catalog # L00441.06
CAS Number:  
Irritant
1-(trans-Cinnamoyl)imidazole 98%
Catalog # L00441.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :198.23 g/mol
Melting Pt:128…132 °C
EINECS:214-510-8
 :GHS07
CAS Number:1138-15-4
Formula:C₁₂H₁₀N₂O
MDL Number:MFCD00005288