
1-(1-Cyclohexen-1-yl)piperidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2981-10-4
:
Formula:C₁₁H₁₉N
MDL Number:MFCD00014643
:165.28 g/mol
Boiling Pt:120…122 °C (16 mmHg)
:GHS07
Related Information
Frequently Bought Together