Order Entry
Northern Ireland
ContactUsLinkComponent
Azithromycin dihydrate
Azithromycin dihydrate
Catalog # J66740.06
CAS Number:  
Azithromycin dihydrate
Catalog # J66740.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance (Color):
    White to off-white
  • Form:
    Powder
  • Assay (unspecified):
    945 µg/mg min.
  • Water Content (Karl Fischer Titration):
    4.0 - 5.0%
  • Optical Rotation:
    -45° to -49°

Specification Test Results

About this item

 :
 :749.00 g/mol
Boiling Pt:717 °C
Melting Pt:126 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Refrigerator
CAS Number:117772-70-0
Formula:C₃₈H₇₂N₂O₁₂·2H₂O
MDL Number:MFCD01862248