Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphamethoxazole
Sulphamethoxazole 98%
Sulphamethoxazole 98%
J66565.22
:
Thermo Fisher Scientific
:
Sulphamethoxazole 98%
J66565.22
:
Thermo Fisher Scientific
:
Pk:
100 g
Seq:
3401
:
:
253.28 g/mol
:
14,8918
:
482 °C
:
168…172 °C
:
>110 °C (230 °F)
:
211-963-3
:
GHS07
:
723-46-6
:
C₁₀H₁₁N₃O₃S
:
MFCD00010546