Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Fluoro-2'-deoxyadenosine
2'-Fluoro-2'-deoxyadenosine 99%
2'-Fluoro-2'-deoxyadenosine 99%
J65441.03
:
Thermo Fisher Scientific
:
2'-Fluoro-2'-deoxyadenosine 99%
J65441.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
64183-27-3
:
:
C₁₀H₁₂FN₅O₃
:
MFCD09750859
:
269.236 g/mol