Order Entry
Northern Ireland
ContactUsLinkComponent
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
Catalog # J65098.03
CAS Number:  
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine 98%
Catalog # J65098.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :687.74 g/mol
Storage Temperature:Refrigerator
Formula:C₃₉H₃₇N₅O₇
MDL Number:MFCD08704196