Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Urocanic acid
cis-Urocanic acid
cis-Urocanic acid
cis-Urocanic acid
Catalog # J64679.MA
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
cis-Urocanic acid
Catalog # J64679.MA
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
Documentation
CAS Number:
7699-35-6
:
Formula:
C₆H₆N₂O₂
MDL Number:
MFCD11045310
Product Details
Documentation