Order Entry
Northern Ireland
ContactUsLinkComponent
 
3,5-Dimethyl-1-adamantanamine hydrochloride
Catalog # J63830.MF
CAS Number:  
3,5-Dimethyl-1-adamantanamine hydrochloride
Catalog # J63830.MF
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

CAS Number:41100-52-1
 :
Formula:C₁₂H₂₁N
MDL Number:MFCD00214336
 :215.77 g/mol
Merck Index:14,5829
Storage Temperature:Ambient
EINECS:255-219-6