
Sodium-4-aminobenzoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:555-06-6
:
Formula:C₇H₇NO₂
MDL Number:MFCD00064395
Melting Pt:>300 °C
:159.12 g/mol
EINECS:209-080-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together