Order Entry
Northern Ireland
ContactUsLinkComponent
Phosphate buffered RIPa with glycerol (2X)
Phosphate buffered RIPa with glycerol (2X)
Catalog # J62939.AK
CAS Number:  
Phosphate buffered RIPa with glycerol (2X)
Catalog # J62939.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Phosphate buffered RIPA with glycerol (2X)
  • Physical form:
    Liquid

Specifications

About this item

Physical Form: Liquid