
Tetraethylene glycol monooctyl ether
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19327-39-0
:
Formula:C₁₆H₃₄O₅
MDL Number:MFCD00043033
:306.443 g/mol
Density:0,969
:GHS07
Related Information
Frequently Bought Together