£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Croton oil
Croton oil
Croton oil
:
Thermo Fisher Scientific
Croton oil
:
Thermo Fisher Scientific
:
8001-28-3
:
:
MFCD00130880
:
74 °C (165 °F)
:
GHS08
:
GHS07
:
Ambient
msds_ALFAJ62367
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Petroleum spirit, 100…120 °C, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
: Merck
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
: Merck
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals