Order Entry
Northern Ireland
ContactUsLinkComponent
Aspartame
Aspartame
Certificates
Aspartame
Certificates

Specifications

  • CAS No.:
    22839-47-0
  • Storage conditions:
    Refrigerator
  • Form:
    Powder
  • MDL No.:
    MFCD00002724
  • Protein synonyms:
    H-Asp-Phe-OMe|(3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid|L-AspartyL-L-phenylalanine methyl ester|N-L-α-Aspartyl-L-phenylalanine 1-Methyl Ester
  • Protein/peptide name:
    Aspartame
  • Purity:
    97,5%
  • Molecular weight:
    294,30 g/mol
  • Molecular formula:
    C₁₄H₁₈N₂O₅

Specifications

Related Information

Frequently Bought Together