β-Nicotinamide adenine dinucleotide reduced disodium salt trihydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:497934-76-6
:
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂·3H₂O
MDL Number:MFCD00150359
:763.46 g/mol (709.41 anhy)
Related Information
Frequently Bought Together