BSA blocking buffer, 3% in TBS
Catalog # J61119.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:250 ml
- Description:BSA blocking buffer, 3% in TBS
- Environmentally Preferable:
- Storage temperature:-30°C to -10°C
- Physical form:Liquid
- Appearance:Yellowish
Specifications
About this item
Storage Temperature: −30 to −10 °C
Physical Form: Liquid
Appearance: Yellowish