Magnesium acetate 1 M in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-72-3
:
Formula:(H₃CCOO)₂Mg
MDL Number:MFCD00003539
:142.39 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together