Order Entry
Northern Ireland
ContactUsLinkComponent
2-Bromo-3-fluoro-6-methylpyridine 98%
Irritant
2-Bromo-3-fluoro-6-methylpyridine 98%
Catalog # H64900.06
CAS Number:  
Irritant
2-Bromo-3-fluoro-6-methylpyridine 98%
Catalog # H64900.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :190.01 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:374633-36-0
Formula:C₆H₅BrFN
MDL Number:MFCD03095061