Order Entry
Northern Ireland
ContactUsLinkComponent
3-[3-(Trifluoromethyl)phenyl]-1-propanol 98%
3-[3-(Trifluoromethyl)phenyl]-1-propanol 98%
Catalog # H64553.MD
CAS Number:  
3-[3-(Trifluoromethyl)phenyl]-1-propanol 98%
Catalog # H64553.MD
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :204.19 g/mol
Density:1,10 g/mL
CAS Number:78573-45-2
Formula:C₁₀H₁₁F₃O
MDL Number:MFCD08706408