Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethyl-4-penten-2-ol
2,4-Dimethyl-4-penten-2-ol ≥97%
2,4-Dimethyl-4-penten-2-ol ≥97%
2,4-Dimethyl-4-penten-2-ol ≥97%
Catalog # B20605.06
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
2,4-Dimethyl-4-penten-2-ol ≥97%
Catalog # B20605.06
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
Documentation
:
UN:
1987
ADR:
,
CAS Number:
19781-53-4
Formula:
C₇H₁₄O
MDL Number:
MFCD00039615
Product Details
Documentation