Order Entry
Northern Ireland
ContactUsLinkComponent
2,6-Dichloro-3-iodopyridine 98%
Irritant
2,6-Dichloro-3-iodopyridine 98%
Catalog # H64366.06
CAS Number:  
Irritant
2,6-Dichloro-3-iodopyridine 98%
Catalog # H64366.06
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:148493-37-2
 :
Formula:C₅H₂Cl₂IN
 :273.88 g/mol
Melting Pt:74…79 °C
ADR:6.1,III
 :GHS07
MDL Number:MFCD03094686