Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
H-D-Asp(OtBu)-OH
H-D-Asp(OtBu)-OH 98%
H-D-Asp(OtBu)-OH 98%
H-D-Asp(OtBu)-OH 98%
Catalog # H63795.06
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Expand image
H-D-Asp(OtBu)-OH 98%
Catalog # H63795.06
Supplier:
Thermo Fisher Scientific
CAS Number:
Certificates
Specifications
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
Documentation
CAS Number:
64960-75-4
:
Formula:
C₈H₁₅NO₄
MDL Number:
MFCD00077101
:
189.211 g/mol
Product Details
Documentation