
L-4-Cyanophenylalanine 98% (by NMR)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:167479-78-9
:
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD00270363
:190.202 g/mol
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together