Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
2-Chloro-1-methylbenzimidazole
Catalog # H63559.03
CAS Number:  
Irritant
2-Chloro-1-methylbenzimidazole
Catalog # H63559.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.608 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1849-02-1
Formula:C₈H₇ClN₂
MDL Number:MFCD00769821