(R)-2-Aminopropanamide hydrochloride 98%
Catalog # H63535.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:71810-97-4
:
Formula:C₃H₉ClN₂O
MDL Number:MFCD00039093
:124.57 g/mol
:GHS07
Storage Temperature:Ambient