

Sodium hexabromoplatinate(IV) hexahydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39277-13-9
:
Formula:Na₂PtBr₆·6H₂O
MDL Number:MFCD00014237
:828.49 g/mol (720.49 anhy)
EINECS:254-398-8
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together