Order Entry
Northern Ireland
ContactUsLinkComponent
N-Boc-sarcosine methyl ester 97%
N-Boc-sarcosine methyl ester 97%
Catalog # H62304.06
CAS Number:  
N-Boc-sarcosine methyl ester 97%
Catalog # H62304.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :203.24 g/mol
CAS Number:42492-57-9
Formula:C₉H₁₇NO₄
MDL Number:MFCD04973115