
1-(4-Biphenylyl)ethylamine 97%
Catalog # H58237.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.28 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:86217-82-5
Formula:C₁₄H₁₅N
MDL Number:MFCD02667819