
Diphenyl-2-pyridylphosphine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37943-90-1
:
Formula:C₁₇H₁₄NP
MDL Number:MFCD00192108
:263.28 g/mol
Melting Pt:82…84 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together