
Isopentyl-4-(dimethylamino)benzoate mixture of isomers ≥99%
Catalog # H55683.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
CAS Number:21245-01-2
:
Formula:C₁₄H₂₁NO₂
MDL Number:MFCD00059356
:235.33 g/mol
Merck Index:14,3232
Boiling Pt:150…155 °C (2,5 mmHg)
Melting Pt:31…36 °C
Density:1,02
EINECS:244-288-8
:GHS07
Storage Temperature:Ambient