
Chlorotris(triphenylphosphine)cobalt(I) 97%
Catalog # H55395.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:881.26 g/mol
Melting Pt:135…139 °C
:GHS07
Storage Temperature:Ambient
CAS Number:26305-75-9
Formula:C₅₄H₄₅ClCoP₃
MDL Number:MFCD00015864