
1-Naphthyltrifluoromethanesulphonate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99747-74-7
:
Formula:C₁₁H₇F₃O₃S
MDL Number:MFCD00192340
:276.229 g/mol
Boiling Pt:97…98 °C (0,3 mmHg)
Density:1,405
Flash Pt:113 °C (235 °F)
Storage Temperature:Ambient
ADR:8,II
:GHS05
Related Information
Frequently Bought Together