

4-(Chlorocarbonyl)benzeneboronic anhydride 90%, tech.
Catalog # H52852.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.369 g/mol
Melting Pt:>300 °C
:GHS06
:GHS07
Storage Temperature:Ambient
CAS Number:332154-57-1
Formula:C₇H₄BClO₂
MDL Number:MFCD06801674