
2-Bromo-3-(bromomethyl)pyridine 96%
Catalog # H52311.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:94446-97-6
:
Formula:C₆H₅Br₂N
MDL Number:MFCD08276299
:250.92 g/mol
Boiling Pt:291 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient