(2S,4R)-Methyl-4-hydroxypyrrolidine-2-carboxylate hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40216-83-9
:
Formula:C₆H₁₁NO₃·HCl
MDL Number:MFCD00080855
:181.6 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together