Order Entry
Northern Ireland
ContactUsLinkComponent
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Irritant
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Catalog # H51050.06
CAS Number:  
Irritant
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Catalog # H51050.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:61478-26-0
 :
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD02094386
 :217.27 g/mol
 :GHS07
Storage Temperature:Ambient