
3-Oxopentanenitrile ≥99%
Catalog # H50789.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:33279-01-5
:
Formula:C₅H₇NO
:97.117 g/mol
Melting Pt:27…30 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
MDL Number:MFCD07367087