Order Entry
Northern Ireland
ContactUsLinkComponent
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole 99%
Irritant
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole 99%
Catalog # H50665.03
CAS Number:  
Irritant
3-(2-Pyridyl)-5-(4-pyridyl)-1,2,4-triazole 99%
Catalog # H50665.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:36770-50-0
 :
Formula:C₁₂H₉N₅
MDL Number:MFCD00107797
 :223.24 g/mol
 :GHS07
Storage Temperature:Ambient