Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-(4-fluorophenyl)nicotinonitrile
2-Chloro-6-(4-fluorophenyl)nicotinonitrile 96%
2-Chloro-6-(4-fluorophenyl)nicotinonitrile 96%
H50519.03
:
Thermo Fisher Scientific
:
2-Chloro-6-(4-fluorophenyl)nicotinonitrile 96%
H50519.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
31776-83-7
:
:
C₁₂H₆ClFN₂
:
MFCD00215072
:
232.65 g/mol
:
GHS07
:
Ambient