Order Entry
Northern Ireland
ContactUsLinkComponent
1-Boc-4-(5-iodo-2-pyridyl)piperazine 95%
Irritant
1-Boc-4-(5-iodo-2-pyridyl)piperazine 95%
Catalog # H50144.03
CAS Number:  
Irritant
1-Boc-4-(5-iodo-2-pyridyl)piperazine 95%
Catalog # H50144.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:497915-42-1
 :
Formula:C₁₄H₂₀IN₃O₂
MDL Number:MFCD07781193
 :389.24 g/mol
 :GHS07
Storage Temperature:Ambient