Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
Irritant
2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
Catalog # H50048.06
CAS Number:  
Irritant
2-Chloro-4-iodopyridine-3-carboxaldehyde 98%
Catalog # H50048.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :267.45 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:153034-90-3
Formula:C₆H₃ClINO
MDL Number:MFCD03095294