
5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-boronic acid 98%
Catalog # H37600.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:169126-63-0
:
Formula:C₁₄H₂₁BO₂
MDL Number:MFCD06801711
:232.13 g/mol
Melting Pt:195…197 °C
:GHS07
Storage Temperature:Ambient